Anti-BCL2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A45148-100UG A45148-50UG
77708-916EA 334.95 USD
77708-916 77708-917
Anti-BCL2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR