Anti-IL17RA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45134-100UG A45134-50UG
77689-169EA 334.95 USD
77689-169 77708-894
Anti-IL17RA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200

Type:
Antigen: IL17RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR