Anti-CCKBR Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A45018-100UG A45018-50UG
77708-685EA 334.95 USD
77708-685 77708-686
Anti-CCKBR Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CCKBR
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR