Anti-ARHGEF5 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44983-100UL
77708-624EA 334.95 USD
77708-624
Anti-ARHGEF5 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:3000

Type:
Antigen: ARHGEF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR