Anti-TTF1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A44669-50UG A44669-100UG
77708-058EA 228.38 USD
77708-058 77708-057
Anti-TTF1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TTF1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR