Anti-COL4A1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A44562-50UG A44562-100UG
77707-869EA 228.38 USD
77707-869 77707-868
Anti-COL4A1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: COL4A1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR