Anti-GRIN2B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44502-100UL
77707-760EA 334.95 USD
77707-760
Anti-GRIN2B Rabbit Polyclonal Antibody
Antibodies
IF:1:100-1:200

Type:
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR