Anti-NRG4 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A44135-100UG A44135-50UG
77707-122EA 334.95 USD
77707-122 77707-123
Anti-NRG4 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NRG4
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR