Anti-NT5E Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A44113-50UL A44113-100UL
77707-094EA 228.38 USD
77707-094 77707-093
Anti-NT5E Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NT5E
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR