Anti-AFAP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43856-50UL A43856-100UL
77706-635EA 228.38 USD
77706-635 77706-634
Anti-AFAP1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:15-1:50

Type:
Antigen: AFAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR