Anti-RGS22 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43552-50UL A43552-100UL
77706-097EA 228.38 USD
77706-097 77706-096
Anti-RGS22 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RGS22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR