Anti-IRX3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43546-100UL
77706-086EA 334.95 USD
77706-086
Anti-IRX3 Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-1:100

Type:
Antigen: IRX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR