Anti-PPP4R1L Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43541-100UL
77706-079EA 334.95 USD
77706-079
Anti-PPP4R1L Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PPP4R1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR