Anti-PPBP Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A43526-100UG A43526-50UG
77706-050EA 334.95 USD
77706-050 77706-051
Anti-PPBP Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PPBP
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR