Anti-PLA2G7 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A43487-50UG A43487-100UG
77705-979EA 228.38 USD
77705-979 77705-978
Anti-PLA2G7 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PLA2G7
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR