Anti-RHEB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43459-50UL A43459-100UL
77705-928EA 228.38 USD
77705-928 77705-927
Anti-RHEB Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RHEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR