SafeBasics™ Beard Covers
Supplier: Medicom
8035
77705-818BG 5.08 USD
77705-818
SafeBasics™ Beard Covers
Disposable Garments
Non sterile white beard covers.
  • Intended to be worn by personnel during various procedures to protect patient/materials
  • Protects from transfer of microorganisms, body fluids, and particulate material
  • Single-use and disposable
  • Latex-free
Highly breathable and low-linting spunbond polypropylene construction.
Packaging: Beard covers are packaged 100 per bag, 10 bags per case (1000 covers per case).