Anti-HMMR Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52433-100UG
77705-498EA 253.75 USD
77705-498
Anti-HMMR Rabbit Polyclonal Antibody
Antibodies
WB:1:500-2000, IHC:1:50-300, ELISA:1:10000-20000

Type:
Antigen: HMMR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR