Anti-Pan myristoylation Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52367-100UG
77705-485EA 253.75 USD
77705-485
Anti-Pan myristoylation Rabbit Polyclonal Antibody
Antibodies
WB:1:500-10000, ELISA:1:10000

Type:
Antigen: Pan myristoylation
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR