Anti-HSP90AA1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43098-100UG
77705-203EA 253.75 USD
77705-203
Anti-HSP90AA1 Rabbit Polyclonal Antibody
Antibodies
WB:1:1000-2000, IHC:1:200-500

Type:
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR