Anti-Mono-Methyl-Histone H3 (Lys18) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43077-100UG
77705-183EA 253.75 USD
77705-183
Anti-Mono-Methyl-Histone H3 (Lys18) Rabbit Polyclonal Antibody
Antibodies
WB:1:200-500

Type:
Antigen: Mono-Methyl-Histone H3 (Lys18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR