Anti-TERF1 (phospho-S219) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40902-100UG
77704-413EA 253.75 USD
77704-413
Anti-TERF1 (phospho-S219) Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TERF1 (phospho-S219)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR