Anti-STK11 (phospho-S334) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40844-100UG
77704-357EA 253.75 USD
77704-357
Anti-STK11 (phospho-S334) Rabbit Polyclonal Antibody
Antibodies
IHC:1:100-1:300, ELISA:1:10000

Type:
Antigen: STK11 (phospho-S334)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR