Anti-DNAI2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40707-100UG
77704-226EA 253.75 USD
77704-226
Anti-DNAI2 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, ELISA:1:10000

Type:
Antigen: DNAI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR