Anti-RAB5C Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40174-100UG
77704-017EA 253.75 USD
77704-017
Anti-RAB5C Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RAB5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR