Anti-PTPRC (phospho-S1007) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A40151-100UG
77703-995EA 253.75 USD
77703-995
Anti-PTPRC (phospho-S1007) Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PTPRC (phospho-S1007)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR