Anti-PRKCG Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39576-100UG
77703-841EA 253.75 USD
77703-841
Anti-PRKCG Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PRKCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR