Anti-NLE1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39456-100UG
77703-721EA 253.75 USD
77703-721
Anti-NLE1 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000

Type:
Antigen: NLE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR