Anti-PKD2 (phospho-S812) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39395-100UG
77703-665EA 253.75 USD
77703-665
Anti-PKD2 (phospho-S812) Rabbit Polyclonal Antibody
Antibodies
IF:1:200-1:1000, ELISA:1:5000

Type:
Antigen: PKD2 (phospho-S812)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR