Anti-PEX1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39369-100UG
77703-639EA 253.75 USD
77703-639
Anti-PEX1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PEX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR