Anti-PPP1R12A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A33425-100UG
77703-403EA 253.75 USD
77703-403
Anti-PPP1R12A Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000

Type:
Antigen: PPP1R12A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR