Anti-MUC1 (phospho-Y1243) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24552-100UG
77703-212EA 253.75 USD
77703-212
Anti-MUC1 (phospho-Y1243) Rabbit Polyclonal Antibody
Antibodies
IHC:1:100-1:300, ELISA:1:20000

Type:
Antigen: MUC1 (phospho-Y1243)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR