Anti-OR52A4P Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A23598-100UG
77702-978EA 253.75 USD
77702-978
Anti-OR52A4P Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: OR52A4P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR