Anti-GRM7 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A22492-100UG
77702-731EA 253.75 USD
77702-731
Anti-GRM7 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:40000

Type:
Antigen: GRM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR