Anti-GNG5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A22341-100UG
77702-624EA 253.75 USD
77702-624
Anti-GNG5 Rabbit Polyclonal Antibody
Antibodies
IF:1:200-1:1000, ELISA:1:20000

Type:
Antigen: GNG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR