Anti-GABRB2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A21782-100UG
77702-485EA 253.75 USD
77702-485
Anti-GABRB2 Rabbit Polyclonal Antibody
Antibodies
IHC:1:100-1:300, ELISA:1:10000

Type:
Antigen: GABRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR