Anti-OR5AS1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A21385-100UG
77702-272EA 253.75 USD
77702-272
Anti-OR5AS1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: OR5AS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR