Anti-FAM117B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A20199-100UG
77701-970EA 253.75 USD
77701-970
Anti-FAM117B Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, ELISA:1:5000

Type:
Antigen: FAM117B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR