Anti-ADRA1D Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A20113-100UG
77701-938EA 253.75 USD
77701-938
Anti-ADRA1D Rabbit Polyclonal Antibody
Antibodies
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000

Type:
Antigen: ADRA1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR