Anti-ITGA2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A18529-100UG
77701-400EA 253.75 USD
77701-400
Anti-ITGA2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: ITGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR