Anti-KRT14 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A16782-100UG
77701-013EA 253.75 USD
77701-013
Anti-KRT14 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: KRT14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR