Anti-LIMK2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A14540-100UG
77699-870EA 253.75 USD
77699-870
Anti-LIMK2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: LIMK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR