Anti-FAS Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A13728-100UG
77699-494EA 253.75 USD
77699-494
Anti-FAS Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:10000

Type:
Antigen: FAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR