Anti-RB1 (Phospho-S807) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A11077-100UG
77698-613EA 253.75 USD
77698-613
Anti-RB1 (Phospho-S807) Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000

Type:
Antigen: RB1 (phospho-S807)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR