Anti-IRS1 (Phospho-S636) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A11014-100UG
77698-554EA 253.75 USD
77698-554
Anti-IRS1 (Phospho-S636) Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: IRS1 (phospho-S636)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR