Anti-Histone H2B (Acetyl-Lys15) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A10896-100UG
77698-445EA 253.75 USD
77698-445
Anti-Histone H2B (Acetyl-Lys15) Rabbit Polyclonal Antibody
Antibodies
WB:1:200-500

Type:
Antigen: Histone H2B (acetyl-Lys15)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR