Anti-EEF1A1 (Acetyl-K41) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A10733-100UG
77698-315EA 253.75 USD
77698-315
Anti-EEF1A1 (Acetyl-K41) Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, ELISA:1:20000

Type:
Antigen: EEF1A1 (acetyl-K41)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR