Anti-COL2A1 Mouse Monoclonal Antibody

Supplier: AFG Bioscience
A10377-100UG
77698-137EA 253.75 USD
77698-137
Anti-COL2A1 Mouse Monoclonal Antibody
Antibodies
Recommended dilution: IHC:1:50-1:200

Type:
Antigen: COL2A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR