Anti-LGALS3 Mouse Monoclonal Antibody

Supplier: AFG Bioscience
A10052-100UG
77697-967EA 253.75 USD
77697-967
Anti-LGALS3 Mouse Monoclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:50-1:500

Type:
Antigen: LGALS3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR