Anti-Plant actin Mouse Monoclonal Antibody

Supplier: AFG Bioscience
A10043-100UG
77697-958EA 253.75 USD
77697-958
Anti-Plant actin Mouse Monoclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: Plant actin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Various Plants
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR